Learn More
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
Brand:  Thermo Scientific™ D50229
Additional Details : Weight : 0.72000kg
 View more versions of this product
Product Code. 10032283
  • € 203.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • Easypure™ II Systems require pretreated feedwater
  • From the chart, select appropriate cartridges based on application and feedwater type
Order Info
Cartridges and filters must be ordered separately
Specifications
Specifications
Reverse Osmosis, Distilled
Ultrapure Mixed Bed
Easypure II Water Purification System, Organic Free/Organic Removal
60 μm
Organicfree/organic Removal
Cartridge and Filter