missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Castaloy™ Three-Prong Extension Clamps
Hold irregular shapes at any angle, surely and safely.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
Sturdy extension clamps allow you to hold objects with irregular shapes at any angle, safely and securely.
- Two separate adjustment screws ensure even, precise pressure on glass and minimize chance of breakage
- Clamps are easily adjustable to vary holding angle, distance from stand
Specifications
1 Ea. | |
Fiberglass and Vinyl sleeve | |
27 cm | |
8.9 cm | |
3-Prong |
Extension Clamp | |
10.5 in. | |
Extension arm O.D.: 0.50 in. (1.3cm) | |
3.5 in. |