Learn More
Thermo Scientific™ CryoExtra™ Racks and Accessories
Product Code. 15152754 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Thermo Scientific CryoExtra 94 Rack
Type:
Cryo Mini Rack
Cryo Square Rack
Cryo Storage Frame
Holds:
2mL, 13 Cardboard Boxes (100-cell Divider)
2mL, 13 Cardboard Boxes (25-cell Divider)
2mL, 15 Cardboard Boxes (100-cell Divider)
2mL, 15 Cardboard Boxes (25-cell Divider)
Fenwal 4R5461 (250 mL Bag), Cassettes (5)
Fenwal 4R5462 (500 mL Bag), Cassettes (5)
Fenwal 4R9951 (50 mL Bag), Cassettes (8)
Unit Size:
Each
Product Code. 15152754
Brand: Thermo Scientific™ 1950685
Only null left
Add to basket
Add to basket
Racks, frames and electrical options
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Mini Rack
For Use With (Equipment) Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Includes 2mL, 13 Cardboard boxes (25 cell divider)
Holds 2mL, 13 Cardboard Boxes (25-cell Divider)