Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Test Tube Holder for 1 in. Tubes
Shop All Thermo Scientific Products
Click to view available options
Type:
Test Tube Holder
Unit Size:
Each
Description
“Snap-in” holder fits 1 in. test tubes up to 104mm tall.
Specifications
Specifications
| Type | Test Tube Holder |
| For Use With (Equipment) | Genesys™ 20 spectrophotometer |
| Holds | One test tube |
| Pathlength | 104 mm |