Learn More
Thermo Scientific™ Nunc™ Caps for TufRol™ Roller Bottles
Click to view available options
Product Type:
Non-vented Cap
Vented Cap
Unit Size:
Pack of 500
Description
Vented and non-vented
- Molded of high density polyethylene (HDPE)
- Designed to prevent misthreading and to provide a secure seal
- Vented cap has an integral hydrophobic 0.2μm microporous membrane filter vent, allowing consistent gas exchange when cap is in the closed position
Specifications
Specifications
| No. per Case | 500 Cs. |
| Sterility | Sterile |
| Product Type | Vented Cap |
| For Use With (Equipment) | TufRol roller bottle |