Learn More
Invitrogen™ RAB11A Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP195
Additional Details : CAS : 69-74-9 Weight : 0.01000kg
Description
RAB11 (residues 654-665) Synthetic Peptide, PEP-195, from Invitrogen.
PEP-195 is a 18 amino acid synthetic peptide. The sequence of this peptide is (amino to carboxy terminus): M G T R D D E Y D Y L F K V V L I C.This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-775. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼ 70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
8766 | |
purified | |
RAB11A | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P62491 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RAB11A | |
No preservative | |
50 μg | |
M G T R D D E Y D Y L F K V V L I C |