missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine Osteocalcin Native Protein
Native Protein
Brand: Invitrogen™ RP43154
Additional Details : Weight : 0.01000kg
Description
Osteocalcin (bovine) Native Protein, RP-43154, from Invitrogen.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium. RP-43154 contains native bovine Osteocalcin protein. RP-43154 may be used as a positive control in many immunological assays. The RP-43154 protein molecular weight is ∽5.8 kDa. The extinction coefficient for a 1% solution of RP-43154 is 13.3.Specifications
P02820 | |
Control | |
5.8 | |
100μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Bovine Osteocalcin | |
Bovine | |
Bv | |
Native | |
Native |
4.9mg/mL | |
50%water with 50% glycerol and no preservative | |
Bovine Osteocalcin | |
Bovine | |
RUO | |
BGLAP | |
Unconjugated | |
Bovine | |
Liquid | |
purified |
in vitro research only.