Learn More
Thermo Scientific™ Pierce™ Premium Grade Sulfo NHS-SS-Biotin
Biotin-label antibodies or other proteins and biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin, a high-quality formulation of this amine-reactive, cleavable biotinylation reagent.
€ 312.00 - € 3260.00
Specifications
Product Type | Sulfo NHS-SS-Biotin |
---|---|
Product Line | Pierce™ |
Cell Permeability | Cell-Impermeant |
Chemical Reactivity | Amine |
Reactive Moiety | Active Ester, Succinimidyl Ester, Sulfo-NHS Ester |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15342627
|
Thermo Scientific™
PG82078 |
1 g |
€ 3260.00
1g |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15332627
|
Thermo Scientific™
PG82077 |
100 mg |
€ 312.00
100mg |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
Thermo Scientific Pierce Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.
- High quality - identity and purity confirmed by multiple tests, including quantitative NMR
- Product integrity - enhanced level of testing and characterization compared to standard grade
- Lot retention - ample supply of past lots retained to help ensure future process testing
- Change management - Change Control Notification (CCN) service
- Consistent manufacture - batch-specific manufacturing documentation review
Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.
Specifications
Sulfo NHS-SS-Biotin | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester | |
Biotin | |
Biotin & Analogs |
Pierce™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length, Cleavable |
For Research Use Only. Not for use in diagnostic procedures.