Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Product Code. 12376962
Encompass_Preferred
Change view
Click to view available options
Material:
Silicone, PTFE
Color:
Blue
Well Shape:
Square
Format:
Flat Underside
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Color Well Shape Format unitSize
12376962 Silicone, PTFE Blue Square Flat Underside Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12376962 Supplier Thermo Scientific™ Supplier No. 14823258
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides superior resealability after up to 100 injections, resists coring and tearing
  • PTFE-coated silicone
  • Eliminates cross contamination of samples
  • Available pre-slit for easy penetration and to reduce vacuum
  • Dry heat autoclavable
  • Superior chemical compatibility
Specifications
Color Blue
Material Silicone, PTFE
Autoclavable Autoclavable
No. of Wells 96
No. per Case 10
Format Flat Underside
Pre-slit Yes
Product Type Sealing Mat
Product Line Webseal
Well Shape Square