Learn More
Thermo Scientific™ Versette ClipTips™
Unique interface locks tip onto Versette automated liquid handling system.
Brand: Thermo Scientific™ 5807
Additional Details : Weight : 0.01000kg
Description
Tips have a clasp or clip that locks in place to ensure an effective seal and will only release when mechanically ejected.
- Unique design – On or Off only – tips lock in place providing a secure fit and preventing loose, uneven or leaky tips
- Minimizing cross contamination between samples, the tips reach the bottom of tubes
- Versatile tip range with three sizes for volumes: 1250μL, 300/125μL and 30/12.5μL
- Rigid rack allows stable, even tip seating
Specifications
100 to 1250 μL | |
Sterile | |
On or off tips lock in place providing a secure fit and preventing loose, uneven or leaky tips, minimizing cross contamination between samples, provide stable, straight mounting every time | |
Automation | |
Versette ClipTips |
Non-Filtered | |
1250μL | |
Rack | |
Thermo Scientific Versette |