Learn More
Karl Hecht™ Thermometer, Red Petrol-Filled
Click to view available options
Product Type:
Dry Thermometer 100
Dry Thermometer 150
Dry Thermometer 250
Dry Thermometer 50
Thermometer
Unit Size:
Each
Description
- Red Petrol-filled for easy readability.
Order Info
ProductTypeSpecs
Specifications
Specifications
| Material | Normal Glass |
| Thermometer Fill | Red petrol |
| Product Type | Dry Thermometer 100 |
| Description | Dry Thermometer 100 |
| Max. Temperature (Metric) | 100°C |
| Temperature Range (Metric) | 50°C |