missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THG1L Polyclonal Antibody

THG1L Polyclonal Antibody
Brand: Invitrogen PA569506
This item is not returnable.
View return policy
Description
THG1L Polyclonal Antibody for Western Blot
Specifications
| THG1L | |
| Polyclonal | |
| Unconjugated | |
| THG1L | |
| hTHG1; ICF45; IHG-1; induced by high glucose-1; interphase cyctoplasmic foci protein 45; interphase cytoplasmic foci protein 45; Probable tRNA(His) guanylyltransferase; Thg1l; tRNA-histidine guanylyltransferase; tRNA-histidine guanylyltransferase 1 like; tRNA-histidine guanylyltransferase 1-like; tRNA-histidine guanylyltransferase 1-like (S. cerevisiae); zgc:101609 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66628 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CY52 | |
| THG1L | |
| synthetic peptide corresponding to a region of mouse THG1L (aa 60-109). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction