Learn More
Thiacloprid, TRC
Product Code. 30133296
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133296 5 g 5g
30041110 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133296 Supplier Toronto Research Chemicals Supplier No. TRCT3439005G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111988-49-9
Molecular Formula C10 H9 Cl N4 S
Molecular Weight (g/mol) 252.72
Synonym Cyanamide, N-[3-[(6-chloro-3-pyridinyl)methyl]-2-thiazolidinylidene]-, [N(Z)]- (ACI), [N(Z)]-N-[3-[(6-Chloro-3-pyridinyl)methyl]-2-thiazolidinylidene]cyanamide (ACI), Cyanamide, [3-[(6-chloro-3-pyridinyl)methyl]-2-thiazolidinylidene]-, [N(Z)]- (9CI), Bariard, Biscaya, Calypso, Calypso (pesticide), Calypso 480SC, Calypso 4F, Calypso 70WG, Calypso Ultra, Sonido, Thiacloprid
IUPAC Name (Z)-[3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene]cyanamide
SMILES Clc1ccc(CN2CCS/C/2=N\C#N)cn1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H9ClN4S/c11-9-2-1-8(5-13-9)6-15-3-4-16-10(15)14-7-12/h1-2,5H,3-4,6H2/b14-10-
Formula Weight 252.0236
Chemical Name or Material Thiacloprid