missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazine Red R, TRC
Specifications
Specifications
| Chemical Name or Material | Thiazine Red R |
| CAS | 2150-33-6 |
| Molecular Formula | C24 H15 N3 O7 S3 . 2 Na |
| Quantity | 100 mg |
| Synonym | Thiazine Red R, 2-[4-[(1-Hydroxy-4-sulfo-2-naphthalenyl)azo]phenyl]-6-methyl-7-benzothiazolesulfonic Acid Disodium Salt, C.I. Direct Red 45, C.I. Direct Red 45','Disodium Salt, 1741 Red, Benzo Red RS, C.I. 14780, C.I. Food Red 13, Chicago Red, Chloramine Red 2BN, Chlorazol Pink Y, Diaphtamine Pink TR, Direct Pink T, Direct Red 45, Direct Thio Red, Hexacol Red FB, Hispamin Pink G, Pontamine Pink 2B, Red FB, Rosophenine 10B, Rosophenine 10B.FQ, Rosophenine G, Thiazine Red R, Tolamine Pink |
| InChI Formula | InChI=1S/C24H17N3O7S3.2Na/c1-13-6-11-18-22(23(13)37(32,33)34)35-24(25-18)14-7-9-15(10-8-14)26-27-19-12-20(36(29,30)31)16-4-2-3-5-17(16)21(19)28;;/h2-12,28H,1H3,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2 |
| SMILES | O=S(C1=C2C(N=C(C3=CC=C(/N=N/C4=C(O)C(C=CC=C5)=C5C(S(=O)(O[Na])=O)=C4)C=C3)S2)=CC=C1C)(O[Na])=O |
| IUPAC Name | disodium;2-[4-[(E)-(1-hydroxy-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]-6-methyl-1,3-benzothiazole-7-sulfonate |
| Molecular Weight (g/mol) | 599.566 |
| Formula Weight | 598.987 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?