Learn More
Thiazole, 99%, Thermo Scientific Chemicals
Catalog Number p-3737553
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
288-47-1
C3H3NS
85.12
MFCD00005315
FZWLAAWBMGSTSO-UHFFFAOYSA-N
thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
9256
CHEBI:43732
1,3-thiazole
S1C=CN=C1
288-47-1
C3H3NS
85.12
MFCD00005315
FZWLAAWBMGSTSO-UHFFFAOYSA-N
thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
9256
CHEBI:43732
1,3-thiazole
S1C=CN=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1980g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5365 to 1.5395
Quantity 1 g
Beilstein 27, 15
Merck Index 15, 9459
Specific Gravity 1.198
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 85.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.