Learn More
Thieno[3,2-b]pyridin-7-ol, 96%
Product Code. p-7033147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419290 1 g 1g
11429290 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11419290 Supplier Thermo Scientific Alfa Aesar Supplier No. H50172.03
Add to basket
CAS: 107818-20-2 | C7H5NOS | 151.183 g/mol
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107818-20-2
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD01863636
InChI Key AACVULYSNJAKEQ-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
PubChem CID 5074765
IUPAC Name 4H-thieno[3,2-b]pyridin-7-one
SMILES C1=CNC2=C(C1=O)SC=C2
Specifications
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 151.19
Percent Purity 96%
Chemical Name or Material Thieno[3,2-b]pyridin-7-ol