missing translation for 'onlineSavingsMsg'
Learn More

Thiirane, TRC

Product Code. 30451292
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451292 1 g 1g
30451228 25 g 25g
30450506 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451292 Supplier Toronto Research Chemicals Supplier No. TRCT3955051G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420-12-2
Molecular Formula C2H4S
Molecular Weight (g/mol) 60.12
Synonym Ethylene Sulfide, Ethylene episulfide, NSC 89690, Thiacyclopropane, 2,3-Dihydro-thiirene
IUPAC Name thiirane
SMILES C1CS1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H4S/c1-2-3-1/h1-2H2
Formula Weight 60.0034
Chemical Name or Material Thiirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.