Learn More
Thiometon Sulfoxide, TRC
Product Code. 30453102
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453102 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30453102 Supplier Toronto Research Chemicals Supplier No. TRCT34467525MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thiometon-sulfoxide
CAS 2703-37-9
Purity Grade Notes HPLC
Molecular Formula C6 H15 O3 P S3
Quantity 25 mg
Synonym Intrathion sulfoxide, Intration sulfoxide, O,O-Dimethyl S-(2-ethylsulfinyl)ethyl dithiophosphate, O,O-Dimethyl S-(2-ethylsulfinylethyl) phosphorodithioate, Oxythiometon, Thiometon sulfoxide, Thiometon-sulfoxide
InChI Formula InChI=1S/C6H15O3PS3/c1-4-13(7)6-5-12-10(11,8-2)9-3/h4-6H2,1-3H3
SMILES CCS(=O)CCSP(=S)(OC)OC
IUPAC Name 2-ethylsulfinylethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 262.35
Formula Weight 261.9921
Percent Purity >95