Learn More
Thionyl chloride, 99.7%, Thermo Scientific Chemicals
Catalog Number p-7214359
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-09-7
Cl2OS
118.96
FYSNRJHAOHDILO-UHFFFAOYSA-N
sulfurooyl dichloride
ClS(Cl)=O
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-09-7
Cl2OS
118.96
FYSNRJHAOHDILO-UHFFFAOYSA-N
sulfurooyl dichloride
ClS(Cl)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -105.0°C
Density 1.6400 g/mL
Boiling Point 76.0°C
Assay Percent Range 99.7%
Packaging Glass bottle
Linear Formula SOCl2
Refractive Index 1.5190 to 1.5210
Quantity 250 mL
Merck Index 15, 9501
Specific Gravity 1.64
Solubility Information Solubility aq. soln.: reacts. Other solubilities: miscible with benzene, chloroform and carbon, tetrachloride
Viscosity 0.6 mPa.s (20°C)
Formula Weight 118.97
Chemical Name or Material Thionyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.