missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-1 Antibody (002), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90730C
This item is not returnable.
View return policy
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Thioredoxin-1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Thioredoxin-1 (P10639; Met1-Val105). | |
| 0.1 mL | |
| Breast Cancer, Lipid and Metabolism | |
| 7295 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 2 | |
| Western Blot, ELISA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction