missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-1 Antibody (116), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89458JF646
This item is not returnable.
View return policy
Description
Thioredoxin-1 Monoclonal antibody specifically detects Thioredoxin-1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Thioredoxin-1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 116 | |
| Western Blot, Immunoprecipitation | |
| TRDX;TRX;TRX1;Trx80;TXN | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Thioredoxin-1 (Uniprot#: P10599; Met1-Val105). | |
| 0.1 mL | |
| Breast Cancer, Lipid and Metabolism | |
| 7295 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction