missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thioredoxin 2 Polyclonal Antibody

Product Code. 16388154
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16388154 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16388154 Supplier Invitrogen™ Supplier No. PA581102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Thioredoxin 2/TXN2.

Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cystein residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide-isomerase (PDI) and a number of transcription factors such as p53, NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thioredoxin 2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TXN2
Gene Accession No. Q99757
Gene Alias 2510006J11Rik; AI788873; COXPD29; G7746; hypothetical protein LOC402938; LMA1; mitochondrial thioredoxin; MTRX; mt-Trx; thioredoxin 2; Thioredoxin II; thioredoxin nuclear gene encoding mitochondrial protein; thioredoxin TRX2; thioredoxin, mitochondrial; Thioredoxin-1; Thioredoxin2; Thioredoxin-2; TR-II; TRX1; Trx2; TRX-2; Txn2; YGR209C; zgc:77127
Gene Symbols TXN2
Host Species Rabbit
Immunogen Recombinant Human Thioredoxin 2/TXN2 Protein (Thr60-Gly166).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25828
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.