missing translation for 'onlineSavingsMsg'
Learn More

Thioredoxin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 140.00 - € 450.00

Specifications

Antigen Thioredoxin
Applications Flow Cytometry
Classification Polyclonal
Conjugate CoraLite 488
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17771259
View Documents
Proteintech
CL488-14999-20UL
20 μL
€ 140.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17639846
View Documents
Proteintech
CL488-14999-100UL
100 μL
€ 450.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thioredoxin Polyclonal antibody apecifically detetects Thioredoxin antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Thioredoxin
Polyclonal
Liquid
Human, Mouse, Rat
BC003377
TXN
IgG
Antigen affinity purification
Flow Cytometry
CoraLite 488
Rabbit
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
7295
Thioredoxin fusion protein Ag6989
Primary
Store at -20°C. Avoid exposure to light.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.