missing translation for 'onlineSavingsMsg'
Learn More

Thioridazine-d3 2-Sulfone, TRC

Product Code. 30449794
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449794 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449794 Supplier Toronto Research Chemicals Supplier No. TRCT3688121MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thioridazine-d3 2-Sulfone
CAS 1329652-09-6
Molecular Formula C21H23D3N2O2S2
Quantity 1 mg
Synonym 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfonyl)-10H-phenothiazine-d3, (+/-)-Thioridazine-2-sulfone-d3, Imagotan-d3, Inofal-d3, Psychoson-d3, Sulforidazine-d3, Sulphoridazine-d3, TPN 12-d3, Thioridazine-2-sulfone-d3
InChI Formula InChI=1S/C21H26N2O2S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)26-21-11-10-17(15-19(21)23)27(2,24)25/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3/i1D3
SMILES CS(C1=CC2=C(C=C1)SC3=CC=CC=C3N2CCC4CCCCN4C([2H])([2H])[2H])(=O)=O
Molecular Weight (g/mol) 405.59
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.