missing translation for 'onlineSavingsMsg'
Learn More

Thiourea, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073609
Click to view available options
Quantity:
250 g
500 g
10 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
62-56-6
CH4N2S
76.12
MFCD00008067
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
2723790
CHEBI:36946
thiourea
NC(N)=S
This item is not returnable. View return policy
62-56-6
CH4N2S
76.12
MFCD00008067
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
2723790
CHEBI:36946
thiourea
NC(N)=S
Melting Point 171°C
Color White
Packaging Plastic powder jar
Quantity 250 g
Formula Weight 76.12g/mol
Physical Form Powder
compliance-icons compliance-icons compliance-icons
Product Identifier
  • THIOUREA SLR
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'