Learn More
MedChemExpress THK5351
Product Code. 16499826
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16499826
Brand: MedChemExpress HY101183
MedChemExpress THK5351 can be radiolabeled and used as a radiotracer for in vivo imaging of tau pathology in the brain.
Specifications
Quantity 50 mg
Product Type Biochemical Assay Reagents
Molecular Weight (g/mol) 327.35 g/mol
Solubility DMSO : 50 mg/mL (152.74 mM; Need ultrasonic)
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
CAS 1707147-26-9