missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THNSL2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26093
This item is not returnable.
View return policy
Description
THNSL2 Polyclonal specifically detects THNSL2 in Human, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| THNSL2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| THNSL2 | |
| Peptide with sequence C-SRFRHREVVHLSR corresponding to internal region according to NP_060741.3, NP_001231605.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_060741.3) and isoform 2 (NP_001231605.1) only. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:128000 | |
| FLJ10916, FLJ35504, secreted osteoclastogenic factor of activated T cells, SOFAT, threonine synthase-like 2 (S. cerevisiae), THS2, TSH2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55258 | |
| Human, Bovine, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction