missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14862-1-AP-20UL
This item is not returnable.
View return policy
Description
Component the THO subcomplex of the TREX complex. The TREX complex specifically associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism. Binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5′ end of the mRNA where it functions in mRNA export. The recruitment occurs via an interaction between THOC4 and the cap-binding protein NCBP1. The TREX complex is essential for the export of Kaposi′s sarcoma-associated herpesvirus (KSHV) intronless mRNAs and infectious virus production. The recruitment of the TREX complex to the intronless viral mRNA occurs via an interaction beteen KSHV ORF57 protein and THOC4. UAP56 functions as a bridge between THOC4 and the THO complex. THOC5 in conjunction with THOC4 functions in NXF1-NXT1 mediated nuclear export of HSP70 mRNA. May be involved in the differentiation of granulocytes and adipocytes.Specifications
| THOC5 | |
| Polyclonal | |
| Unconjugated | |
| THOC5 | |
| C22orf19, Fmip, fSAP79, hTREX90, KIAA0983, PK1.3, Placental protein 39.2, PP39.2, THO complex 5, THO complex subunit 5 homolog, THOC5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107829, 360972, 8563 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13769, Q68FX7, Q8BKT7 | |
| Thoc5 | |
| THOC5 Fusion Protein Ag6664 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction