Learn More
Novus Biologicals™ THOC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30310
This item is not returnable.
View return policy
Description
THOC7 Polyclonal antibody specifically detects THOC7 in Human,Mouse samples. It is validated for Western Blot
Specifications
| THOC7 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| FLJ23445, fSAP24NIF3L1-binding protein 1, Functional spliceosome-associated protein 24, Ngg1 interacting factor 3 like 1 binding protein 1, Ngg1-interacting factor 3-like protein 1-binding protein 1, NIF3L1BP1hTREX30, THO complex 7 homolog (Drosophila), THO complex subunit 7 homolog | |
| Between 1 and 50 | |
| 100 μg | |
| Epigenetics | |
| 80145 | |
| Store at 4°C. Do not freeze. | |
| IgG |