missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOP1 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501409
This item is not returnable.
View return policy
Description
Involved in the metabolism of neuropeptides under 20 amino acid residues long. Involved in cytoplasmic peptide degradation. Able to degrade the beta-amyloid precursor protein and generate amyloidogenic fragments.Specifications
| THOP1 | |
| Monoclonal | |
| 0.67 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52888 | |
| THOP1 | |
| Full length human recombit protein of human THOP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D3 | |
| Unconjugated | |
| THOP1 | |
| EP24.15, MEPD_HUMAN, MP78, TOP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7064 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction