missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPBPL Monoclonal Antibody (OTI5D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505551
This item is not returnable.
View return policy
Description
Tapasin, or TAPBP, is a member of the variable-constant Ig superfamily that links major histocompatibility complex (MHC) class I molecules to the transporter associated with antigen processing in the endoplasmic reticulum (ER). The TAPBP gene is located near the MHC complex on chromosome 6p21.3. TAPBPL is a member of the Ig superfamily that is localized on chromosome 12p13.3, a region somewhat paralogous to the MHC.Specifications
| TAPBPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BX59 | |
| TAPBPL | |
| Full length human recombit protein of human TAPBPL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D7 | |
| Unconjugated | |
| TAPBPL | |
| TAPBP-R, TAPBPR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction