missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRA Monoclonal Antibody (OTI6B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805056S
This item is not returnable.
View return policy
Description
THRA Monoclonal antibody specifically detects THRA in Human samples. It is validated for Western BlotSpecifications
| THRA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13133 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10062 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6B7 | |
| Unconjugated | |
| NR1H3 | |
| 6430529J03Rik; AR7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; AW259572; cerbA protein; c-erb-A protein; c-erb-A thyroid hormone receptor; c-ERBA-1; cerbAalpha; c-erbAalpha; c-erbA-alpha; c-erbAalpha2; c-erbA-alpha2; c-erbAalpha2; thyroid hormone receptor alpha; CHNG6; E ERB-T-1; EAR7; EAR-7; ERBA; ERBA ALPHA; ERBA related 7; ERBA1; ERBA-related 7; ERB-T-1; MGC000261; MGC43240; NR1A1; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Rvr; T3R[a]; T3Ralpha; Thra; THRA1; THRA2; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor alpha 2; thyroid hormone receptor beta; thyroid hormone receptor, alpha; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyron | |
| Human recombinant protein fragment corresponding to amino acids 69-329 of human NR1H3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction