missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRA Mouse anti-Human, Clone: OTI2A2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805187
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| THRA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10827 | |
| THRA | |
| Human recombit protein fragment corresponding to amino acids 1-220 of human THRA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A2 | |
| Unconjugated | |
| THRA | |
| AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7067 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction