missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRA, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00007067-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MEQKPSKVECGSDPEENSARSPDGKRKRKNGQCSLKTSMSGYIPSYLDKDEQCVVCGDKATGYHYRCITCEGCKGFFRRTIQKNLHPTYSCKYDSCCVIDKITRNQCQLCRFKKCIAVGMAMDLVLDDSKRVAKRKLIEQNRERRRKEEMIRSLQQRPEPTPEEWDLIHIATEAHRSTNAQGSHWKQRRKFLPDDIGQSPIVSMPDGDKVDLEAFSEFTKIITPAITRVVDFAKKLPMFSELPCEDQIILLKGCCMEIMSLRAAVRYDPESDTLTLSGEMAVKREQLKNGGLGVVSDAIFELGKSLSAFNLDDTEVALLQAVLLMSTDRSGLLCVDKIEKSQEAYLLAFEHYVNHRKHNIPHFWPKLLMKVTDLRMIGACHASRFLHMKVECPTELFPPLFLEVFEDQEVSpecifications
| THRA | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_199334.2 | |
| THRA | |
| THRA (NP_955366.1, 1 a.a. ∼ 410 a.a) full-length human protein. | |
| RUO | |
| 7067 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian) | |
| AR7/EAR7/ERB-T-1/ERBA/ERBA1/MGC000261/MGC43240/NR1A1/THRA1/THRA2/c-ERBA-1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction