missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THRAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101073
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MED24.
This gene encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| THRAP4 | |
| Polyclonal | |
| Unconjugated | |
| MED24 | |
| 911GSE; activator-recruited cofactor 100 kDa component; ARC100; AU040102; AW547152; Cofactor required for Sp1 transcriptional activation subunit 4; cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa; CRSP complex subunit 4; CRSP100; CRSP4; D11Ertd307e; DRIP100; genetic suppressor element 2; Gse2; hTRAP100; KIAA0130; MED24; MED5; Mediator complex subunit 24; mediator of RNA polymerase II transcription subunit 24; mediator of RNA polymerase II transcription, subunit 24 homolog; MGC8748; mTRAP100; peroxisome proliferation activation receptor binding protein 2; Pparb2; Pparbp2; R75526; THRAP4; thyroid hormone receptor associated protein 4; thyroid hormone receptor-associated protein 100 kDa; thyroid hormone receptor-associated protein 4; Thyroid hormone receptor-associated protein complex 100 kDa component; TRAP100; Vitamin D3 receptor-interacting protein complex 100 kDa component; vitamin D3 receptor-interacting protein complex component DRIP100 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23989, 619436, 9862 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75448, Q4V8B3, Q99K74 | |
| MED24 | |
| A synthesized peptide derived from human MED24(Accession O75448), corresponding to amino acid residues V814-N864. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction