Learn More
SCAT™ Polypropylene Thread Adapters
Product Code. 10130180 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Size:
Thread 1: GL/S40 (f), Thread 2: GL45 (m)
Thread 1: S 60/61 (f) Thread 2: GL45 (m)
Thread 1: S51 (f), Thread 2: GL45 (m)
Thread 1: S55 (f), Thread 2: GL45 (m)
Thread 1: S60/61, Thread 2: S55 (m)
Closure Color:
Colorless
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Closure Color unitSize
10130180 Thread 1: S51 (f), Thread 2: GL45 (m) Colorless Each
10795821 Thread 1: GL/S40 (f), Thread 2: GL45 (m) Colorless Each
10508130 Thread 1: S55 (f), Thread 2: GL45 (m) Colorless Each
10211750 Thread 1: S 60/61 (f) Thread 2: GL45 (m) Colorless Each
10323870 Thread 1: S60/61, Thread 2: S55 (m) Colorless Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10130180 Supplier SCAT™ Supplier No. 107093
Only null left
Add to basket
Add to basket
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Closure Size Thread 1: S51 (f), Thread 2: GL45 (m)
Closure Color Colorless
Liner Material Polypropylene
Product Type Thread Adapter