Learn More
Cole-Parmer™ Threaded Reducing Nipples
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-28
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11878043
Quantity Price
1 € 32.64 / Each
10 € 29.63 / Each
Estimated Shipment: 12-08-2021
Add to basket
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Translucent
Autoclavable
Threaded Male Reducer
1/2 in.
1/2 in. x 3/8 in. NPT(M)
NPT(M) Connection
PFA
3/8 in.
Autoclave or Ethylene Oxide