Learn More
Lenz Laborglasintrumente Three-Neck Round-Bottom Flasks Side Necks Parallel
Product Code. p-7222470
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
4 L
5 L
6 L
10 L
Diameter (Metric) Outer:
131 mm
166 mm
185 mm
207 mm
223 mm
236 mm
279 mm
Standard Taper Joint:
Center Neck NS 29/32, Side Necks NS 14/23
Center Neck NS 29/32, Side Necks NS 19/26
Center Neck NS 29/32, Side Necks NS 29/32
Center Neck NS 45/40, Side Necks NS 29/32
Unit Size:
Each
Product Code. 10498782
Brand: Lenz Laborglasintrumente 03304270
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Reaction flasks acc. to DIN 12392
  • With two parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 1 L
Material Duran Glass
Standard Taper Joint Center Neck NS 29/32, Side Necks NS 19/26
Diameter (Metric) Outer 131 mm