Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
Product Code. 15804538 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24, 29.2 NS
Unit Size:
Pack of 100
Product Code. 15804538
Brand: DWK Life Sciences 286046107
Only null left
Add to basket
Add to basket
For securing the hollow DURAN™ tap plug in the ground DURAN™ single way stopcock.
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
  • 3-part, for hollow tap plugs
  • Three part retaining device holds and secures the tap plug in the tap sleeve
TRUSTED_SUSTAINABILITY
Spécification
Material Borosilicate Glass (3.3), Polypropylene
Autoclavable Autoclavable
Closure Size 24, 29.2 NS
No. per Pack 100/Pk.
For Use With (Application) For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap