Learn More
Buerkle™ PTFE Valves
Product Code. p-8003096
Click to view available options
For Use With:
10 mm Tube
4.5 mm Tube
6.8 mm Tube
Product Type:
2-Way Valve
3-Way Valve
Unit Size:
Each
Product Code. 10257232
Brand: Buerkle™ 86100006
Only null left
Add to basket
Add to basket
Make tubing connections with Buerkle™ Valves. They are autoclavable and they can accommodate varying diameters of tubing sections and configurations.
  • Made of solid PTFE with hose nozzles
  • Resistant to chemicals
  • Very easy connection thanks to its excellent antifriction properties
  • Blue handles made of PP
  • Resistant to compression max. 1 bar
  • Vacuum suitable up to 10 ¯¹ mmHg
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Polypropylene, PTFE
For Use With 10 mm Tube
Width (Metric) 4 mm
Color Blue Handle
Product Type 3-Way Valve