missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombin-antithrombin (TAT) complexes Antibody (22), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41381AF532
This item is not returnable.
View return policy
Description
Thrombin-antithrombin (TAT) complexes Monoclonal antibody specifically detects Thrombin-antithrombin (TAT) complexes in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Thrombin-antithrombin (TAT) complexes | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| F2 | |
| Thrombin-antithrombin (TAT) complexes | |
| Primary | |
| Antibody is specific for human Thrombin-antithrombin (TAT) complexes. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry | |
| 22 | |
| Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA | |
| Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
| Mouse | |
| 0.1 mL | |
| 2147 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction