missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (461714), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3894PECY7
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 461714 | |
| Flow Cytometry | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Mouse myeloma cell line NS0-derived recombinant mouse Thrombomodulin/BDCA-3, Leu17-Ser517, Accession # P15306 | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction