missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42213AF700
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Mouse | |
| 60.3 kDa | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4C in the dark. | |
| IgG1 λ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6C8.2F9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| P07204 | |
| THBD | |
| Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction