missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42213
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 4-5 μg/mL, Flow Cytometry 1 μg/ million cells, Immunohistochemistry 10 - 15 μg/mL, Immunohistochemistry-Paraffin 10 - 15 μg/mL | |
| P07204 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6C8.2F9 | |
| Unconjugated | |
| PBS | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
| 0.1 mg | |
| Cardiovascular Biology | |
| 7056 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction