missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (THBD/8188R), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21031DL594
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| DyLight 594 | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Recombinant full-length human Thrombomodulin/BDCA-3 protein | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| THBD/8188R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction