Learn More
Thrombomodulin / CD141 Mouse, Clone: THBD/1591, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7056-MSM1-P0
This item is not returnable.
View return policy
Specifications
| Thrombomodulin / CD141 | |
| Cell Surface and Cytoplasmic | |
| THBD/1591 | |
| Unconjugated | |
| NP_000352 | |
| Mouse | |
| 100kDa | |
| Protein A/G Purified | |
| RUO | |
| 7056 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| THBD | |
| Recombinant human Thrombomodulin protein fragment (aa69-194) | |
| THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |