missing translation for 'onlineSavingsMsg'
Learn More

Thrombomodulin / CD141 Mouse, Clone: THBD/1782, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 7056-MSM2-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940968

  • € 462.00 / 100µg

Estimated Shipment: 10-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Thrombomodulin / CD141
Cell Surface and Cytoplasmic
THBD/1782
Unconjugated
NP_000352
Mouse
100kDa
Protein A/G Purified
RUO
7056
IgG2a κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
THBD
Recombinant human Thrombomodulin protein fragment (aa69-194)
THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers