Learn More
Thrombopoietin R/Tpo R Antibody (S03-1C9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19792-100ul
This item is not returnable.
View return policy
Description
Thrombopoietin R/Tpo R Monoclonal antibody specifically detects Thrombopoietin R/Tpo R in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Thrombopoietin R/Tpo R | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-1C9 | |
| Unconjugated | |
| CD110, CD110 antigen, C-MPL, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TPO-R, TPORMPLV | |
| A synthetic peptide of human Thrombopoietin R/Tpo R (Uniprot # P40238) | |
| 100 μg | |
| CD Markers, Immunology, Stem Cells | |
| 4352 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |