missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Thrombopoietin R/Tpo R Antibody (TA136) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | Thrombopoietin R/Tpo R |
|---|---|
| Clone | TA136 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045048
|
Novus Biologicals™
NBP3-28372-100ug |
100 μg |
€ 337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047938
|
Novus Biologicals™
NBP3-28372-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045589
|
Novus Biologicals™
NBP3-28372-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thrombopoietin R/Tpo R Monoclonal antibody specifically detects Thrombopoietin R/Tpo R in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Thrombopoietin R/Tpo R | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| CD Markers, Immunology, Stem Cells | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD110, CD110 antigen, C-MPL, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TPO-R, TPORMPLV | |
| MPL/ TPOR/ CD110 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| TA136 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P40238 | |
| 4352 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title