missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin 1 Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA511330
This item is not returnable.
View return policy
Description
MA5-11330 targets Thrombospondin in WB applications and shows reactivity with Human samples. The MA5-11330 immunogen is purified human TSP.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Specifications
| Thrombospondin 1 | |
| Cocktail | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P07996 | |
| THBS1 | |
| Purified human TSP | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| D4.6, A6.1, MBC 200.1 | |
| Unconjugated | |
| THBS1 | |
| glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7057 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction